missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

£25.00 - £276.00

Chemical Identifiers

CAS 193978-23-3
Molecular Formula C10H15BO2S
Molecular Weight (g/mol) 210.098
MDL Number MFCD05663878
InChI Key FFZHICFAHSDFKZ-UHFFFAOYSA-N
Synonym 2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
PubChem CID 10703628
IUPAC Name 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15430719
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63819.03
1 g
£25.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15440719
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63819.06
5 g
£71.70
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15450719
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63819.14
25 g
£276.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

193978-23-3
210.098
FFZHICFAHSDFKZ-UHFFFAOYSA-N
10703628
B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2
C10H15BO2S
MFCD05663878
2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane

Specifications

193978-23-3
C10H15BO2S
2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2
210.098
228.12
Solid
65°C to 70°C
MFCD05663878
FFZHICFAHSDFKZ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
10703628
98%
Thiophene-2-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

missing translation for 'storageNote' : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only