missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene-2-carbonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Density | 1.171 |
Boiling Point | 191°C to 193°C |
Flash Point | 81°C (178°F) |
Odor | Unpleasant |
Refractive Index | 1.5645 |
Quantity | 25 g |
UN Number | UN3276 |
Beilstein | 107035 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 109.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'