Learn More
Thiophene-2-carboxylic acid, 99%, Thermo Scientific Chemicals
£15.10 - £143.00
Chemical Identifiers
CAS | 527-72-0 |
---|---|
Molecular Formula | C5H4O2S |
Molecular Weight (g/mol) | 128.15 |
MDL Number | MFCD00005437 |
InChI Key | QERYCTSHXKAMIS-UHFFFAOYSA-N |
Synonym | 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid |
PubChem CID | 10700 |
ChEBI | CHEBI:71241 |
IUPAC Name | thiophene-2-carboxylic acid |
SMILES | OC(=O)C1=CC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464164
|
Thermo Scientific Alfa Aesar
A12514.14 |
25 g |
£15.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474164
|
Thermo Scientific Alfa Aesar
A12514.22 |
100 g |
£35.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484164
|
Thermo Scientific Alfa Aesar
A12514.36 |
500 g |
£143.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A hypoglycemic, anti-lipolytic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
527-72-0 | |
128.15 | |
QERYCTSHXKAMIS-UHFFFAOYSA-N | |
10700 | |
thiophene-2-carboxylic acid |
C5H4O2S | |
MFCD00005437 | |
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
CHEBI:71241 | |
OC(=O)C1=CC=CS1 |
Specifications
527-72-0 | |
1.42 | |
C5H4O2S | |
110150 | |
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
OC(=O)C1=CC=CS1 | |
128.15 | |
CHEBI:71241 | |
99% |
126°C to 130°C | |
259°C to 261°C | |
MFCD00005437 | |
14,9354 | |
QERYCTSHXKAMIS-UHFFFAOYSA-N | |
thiophene-2-carboxylic acid | |
10700 | |
128.15 | |
Thiophene-2-carboxylic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
missing translation for 'einecsNumber' : 208-423-4
missing translation for 'rtecsNumber' : XM8330200
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only