missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99+%, extra pure, Thermo Scientific Chemicals

£18.19 - £159.80

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10123380
View Documents Promotion Details
Thermo Scientific Acros
138821000
100 g Glass bottle
£18.19
100g
Please to purchase this item. Need a web account? Register with us today!
10316880
View Documents
Thermo Scientific Acros
138825000
500 g Glass bottle
£50.35
500g
Please to purchase this item. Need a web account? Register with us today!
10444405
View Documents
Thermo Scientific Acros
138820025
2.5 kg Glass bottle
£159.80
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-02-1
84.14
YTPLMLYBLZKORZ-UHFFFAOYSA-N
8030
thiophene
C4H4S
MFCD00005413
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
CHEBI:30856
S1C=CC=C1

Specifications

110-02-1
1.0650g/mL
−9°C
99% min. (GC)
1.5270 to 1.5290
17, 29
1.065
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
S1C=CC=C1
84.14
CHEBI:30856
99+%
Liquid
-38.0°C
84.0°C
Authentic
C4H4S
MFCD00005413
15, 9506
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiophene
8030
84.14
Extra Pure
Thiophene, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-729-4

missing translation for 'rtecsNumber' : XM7350000

missing translation for 'tsca' : TSCA

Product Identifier
  • Thiophene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only