missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, 99%, Thermo Scientific Chemicals

Product Code. 10715631
Click to view available options
Quantity:
25 mL
500 mL
1 L
5 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-98-5
MDL Number MFCD00004826
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
PubChem CID 7969
ChEBI CHEBI:48498
IUPAC Name benzenethiol
SMILES C1=CC=C(C=C1)S

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 1.0780g/mL
Boiling Point 167.0°C to 169.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H6S
Linear Formula C6H5SH
Refractive Index 1.5880 to 1.5900
Quantity 1 L
Beilstein 06, 294
Fieser 06,585; 07,367; 09,465; 10,399; 12,485; 13,53; 16,327
Merck Index 15, 9508
Specific Gravity 1.078
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2
Molecular Weight (g/mol) 110.17
Viscosity 0.981 mm2/s (30°C)
Formula Weight 110.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'