missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiosulphate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£46.55 - £96.60
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12942215
|
Fisher Chemical
A 049 100 |
100 mL | Bottle |
£46.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12952215
|
Fisher Chemical
A 049 250 |
250 mL | Bottle |
£66.35
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12962215
|
Fisher Chemical
A 049 500 |
500 mL | Bottle |
£96.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
S2O32 |
Specifications
1g/L | |
S2O32 | |
Liquid |
Single-component Standard Solution | |
H2O | |
Thiosulphate |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)