Learn More
Thiourea, 99%, Thermo Scientific Chemicals
Description
Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.
Solubility
Soluble in water.
Notes
Stable. Incompatible with strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas.

Specifications
Specifications
Melting Point | 172°C to 178°C |
Density | 1.405 |
Odor | Odorless |
Quantity | 500 g |
UN Number | UN3077 |
Beilstein | 605327 |
Merck Index | 14,9367 |
Solubility Information | Soluble in water. |
Formula Weight | 76.12 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252824 | Certificate of Analysis | 26/11/2024 | A12828.22,A12828.0E,A12828.0C,A12828.36 |
10251766 | Certificate of Analysis | 30/08/2024 | A12828.22,A12828.0E,A12828.0C,A12828.36 |
10229782 | Certificate of Analysis | 04/01/2024 | A12828.22,A12828.0E,A12828.0C,A12828.36 |
M25F042 | Certificate of Analysis | 22/11/2021 | A12828.22,A12828.0E,A12828.0C,A12828.36 |
A15T054 | Certificate of Analysis | 22/11/2021 | A12828.22,A12828.0E,A12828.0C,A12828.36 |
Safety and Handling



- Thiourea
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Thiourea
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.