missing translation for 'onlineSavingsMsg'
Learn More

Thiourea, ACS, 99% min.

Product Code. 11378048
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11378048

Brand: Thermo Scientific Alfa Aesar 036609.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for organic synthesis | CAS: 62-56-6 | CH4N2S | 76.12 g/mol

Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 175°C to 178°C
Color White
Density 1.405 g/mL
Odor Odorless
Linear Formula H2NCSNH2
Quantity 1 kg
UN Number UN3077
Beilstein 605327
Merck Index 14,9367
Solubility Information Soluble in water.
Formula Weight 76.12
Percent Purity ≤99%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Thiourea
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.