Learn More
Mettler Toledo™ Thiourea Cleaning Solution
Removes silver sulfide contamination from pH electrodes. Mettler Toledo™ Thiourea Cleaning Solution helps maintain electrode performance and should be utilized after measurements and prior to recalibration. Treatment should be applied until discoloration.
Brand: Mettler Toledo™ 51350102
31.89 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
The pH electrode is the main actor of pH measurement. It requires special care and attention. The removal of contamination is required before the electrode can be re-calibrated and used for another measurement. Rinsing with deionized water is not sufficient when silver sulfide contamination is present; Thiourea is recommended in this case.
- Available in a handy screw-cap bottle with a no-fuss label
- SDS and label contain information according to GHS (Globally Harmonized System)
- Treatment time is approximately 1 hour

Specifications
Colorless | |
Cleaning solution Thiourea, 250mL | |
Cleaning | |
Odorless | |
1 | |
Completely miscible in water |
Thiourea/thiocarbamide (5–10%) and HCl | |
pH Electrode | |
For Cleaning Junctions with Silver Sulfide Contamination | |
Bottle | |
250 mL |
Safety and Handling
- Cleaning solution Thiourea / HCl
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : thiourea, Hydrochloric acid
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.