missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiourea, Cytiva
Click to view available options
Quantity:
100 g
Specifications
CAS | 45-001-296 |
Color | White |
Molecular Formula | CH4N2S |
Quantity | 100 g |
Formula Weight | 76.13 |
Physical Form | Solid |
Chemical Name or Material | Thiourea |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P235-Keep cool.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'