missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ THIQ

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Potent and selective melanocortin 4 (MC4) receptor agonist (IC50 values are 1.2, 761 and 2067 nM for human MC4, MC3 and MC1 receptors respectively). Enhances intracavernosal pressure and stimulates erectile activity in rats ex copula following systemic administration.
Specifications
Specifications
| CAS | 312637-48-2 |
| Quantity | 1 mg |
| Product Type | THIQ |
| Purity | >95% |
| Molecular Weight (g/mol) | 589.181 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction