missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Counting Chambers, Thoma pattern, BLAUBRAND™, double ruling
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
Brand: Brand™ 718020
55.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
- Rulings correspond to the central large square of the Neubauer chamber.
- The mini squares have an area of 0.0025 mm2 each.
- Since the outer large squares are not completed, the Thoma chamber is only used for counting thrombocytes and erythrocytes.
Specifications
Counting Device | |
0.1mm | |
With spring clips |
1Each | |
Counting thrombocytes and erythrocytes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brand™ Counting Chambers, Thoma pattern, BLAUBRAND™, double ruling > With Spring Clips
Spot an opportunity for improvement?Share a Content Correction