Learn More
Euro Protection™ Thor Multirisk Pants, Navy
Click to view available options
Material:
Cotton, Polyester
Product Type:
Multirisk Pants
Resistant To:
Fire-retardant, Anti-static
Certifications/Compliance (Europe):
EN ISO 11611:2007Class 1 - A1IEC 61482-2:2009 Class 1 - 4 kAEN 13034:2005+A1:2009Type PB6EN 1149-5:200N 14404:2004+A1:2010Type 2
Style:
Includes reflective band tape
Description
- Fire retardant
- Antistatic
- Reflective band tape
- Cotton/polyester construction
Specifications
Specifications
Size | 3X-Large |
Product Type | Multirisk Pants |
Material | Cotton, Polyester |
Resistant To | Fire-retardant, Anti-static |
Certifications/Compliance (Europe) | EN ISO 11611:2007Class 1 - A1IEC 61482-2:2009 Class 1 - 4 kAEN 13034:2005+A1:2009Type PB6EN 1149-5:200N 14404:2004+A1:2010Type 2 |
Style | Includes reflective band tape |