missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ Angle Adapter
Click to view available options
Closure Size:
S60/61
Unit Size:
Each
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Closure Size | S60/61 |
| Closure Color | Black |
| Description | Internal S60 Female to External S60 Male Thread |
| Color | Black |
| Liner Material | HDPE-EC |
| For Use With (Equipment) | Electroconductive Container |
| Material | HDPE-EC |
| Product Type | Thread Adapter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction