missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Thread Adapter

Product Code. p-7084245
Click to view available options
Closure Size:
GL14, NPT 1/4 in.
GL25, 14.6 mm
S60/61, 2 in. Tri-Sure
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10811561

Brand: SymLine™ 107082

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.

SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.

  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards

Specifications

Closure Size S60/61, 2 in. Tri-Sure
Closure Color Black
Description S60/61 (f) to 2 in. Tri-sure (f)
Color Black
Liner Material HDPE-EC
Material HDPE-EC
Product Type Thread Adapter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.