Learn More
Buerkle™ Thread Adapter for Mini Solvent Pump
Thread adapter for Buerkle™ Mini solvent pump.
Brand:  Buerkle™ 0006-4000
41.94 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 15362255
  • £44.15 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Buerkle Mini Solvent Pump
  • Made of ETFE or PTFE
  • External thread: GL 45
Specifications
Specifications
Mini Solvent Pump
PTFE
GL45-S40
Thread Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Thread Adapter for Mini Solvent Pump > External Thread: GL 45; Internal Thread:S 40; PTFE