Learn More
Buerkle™ Thread Adapter for Mini Solvent Pump
Thread adapter for Buerkle™ Mini solvent pump.
Brand:  Buerkle™ 0006-4002
 View more versions of this product
Product Code. 15382255
  • £41.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Buerkle Mini Solvent Pump
  • ETFE Construction
  • External thread: GL 45
Specifications
Specifications
Transparent
ETFE
GL45-GL38
0.021 kg
Mini Solvent Pump
GL45-GL38
Thread Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Thread Adapter for Mini Solvent Pump > External Thread: GL 45; Internal Thread: GL 38, ETFE