Learn More
SCAT™ Thread Adapter, Type 71
Shop All SCAT Europe Products
Click to view available options
Closure Size:
GL38, GL45
Unit Size:
Each
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
| Closure Size | GL38, GL45 |
| Closure Color | Colorless |
| Liner Material | Polypropylene |
| Product Type | Thread Adapter |