Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand: SCAT™ 107994
44.66 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Thread 1: GL/S40 (f), Thread 2: GL45 (m) | |
Colorless | |
Polypropylene | |
Thread Adapter |
Product Content Correction
Product Title
SCAT™ Polypropylene Thread Adapters > Thread Adapters; Closure Size: Thread 1: GL/S40 (f), Thread 2: GL45 (m); Color: Colorless
Spot an opportunity for improvement?Share a Content Correction