Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand:  SCAT™ 108081
41.45 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 14716301
  • £55.70 / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
Thread 1: GL45 (m), Thread 2: S40/41 (f)
Colorless
Polypropylene
Thread Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCAT™ Polypropylene Thread Adapters > Thread Adapters; Closure Size: Thread 1: GL45 (m), Thread 2: S40/41 (f); Color: Colorless