Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand:  SCAT™ 107146
63.21 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.30700kg
 View more versions of this product
Product Code. 17987240
  • £77.36 / Each
Estimated Shipment: 06-08-2024
to see stock.
Description
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
Thread 1: GL45 (m), Thread 2: S65 (f)
Gray
Thread Adapter