Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand:  SCAT™ 108325
161.88 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.94700kg
 View more versions of this product
Product Code. 17917270
  • £198.10 / Each
Estimated Shipment: 06-08-2024
to see stock.
Description
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polytetrafluoroethylene (PTFE) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
S60/61 (m) to B83 (f)
PTFE
Thread Adapter