Learn More
Masterflex™ Threaded Barb Adapter Kits, PVDF
Organize the fittings at your workbench with these convenient fittings kits
Brand:  Masterflex™ 31514-36
Product Code. 11770429
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Designed for quick and easy assembly without tools.
  • Combine these simple and economical fittings with flexible tubing.
  • For use in low-pressure applications (generally less than 30psi).
Specifications
Specifications
Threaded Barb Adapter Kit
Barb x NPT(M), Large
PVDF
1/Pk.