missing translation for 'onlineSavingsMsg'
Learn More

Idex Threaded Luer PEEK Adapter

Product Code. p-4911080
Click to view available options
Color:
Black
Natural
For Use With (Equipment):
Female Luer x Female M6 Flat Bottom
Male Luer Lock x Female 10-32 Coned
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12947759

Brand: Idex 0202359

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Convenience of quick connect and disconnect of a luer connection

  • Manufactured from inert polymers

Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.

Specifications

Type Threaded Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 125°C
Material PEEK
Color Black
For Use With (Equipment) Female Luer x Female M6 Flat Bottom
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.