Learn More
Masterflex™ Threaded Male Adapter Elbow
Designed for quick and easy assembly without tools
Marca: Masterflex™ 31702-28
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Adapts from a NPT male thread to a hose barb.
- Combine these simple and economical fittings with our flexible tubing for use in low-pressure applications (generally less than 30psi).
- Barb fittings are available in a variety of sizes, materials, and configurations from straight connectors to threaded adapter elbows to branched tees.
- A free certificate of resin compliance is available for many formulations.
- Available in a variety of materials and configurations.
Especificaciones
Threaded Adapter | |
Nylon | |
Ethylene Oxide |
Barb x NPT(M) | |
10/Pk. |