Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
For securing the hollow DURAN™ tap plug in the ground DURAN™ single way stopcock.
Brand: DWK Life Sciences 286044209
This item is not returnable.
View return policy
Description
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
- 3-part, for hollow tap plugs
- Three part retaining device holds and secures the tap plug in the tap sleeve
Specifications
Borosilicate Glass (3.3), Polypropylene | |
18.8 NS | |
For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap |
Autoclavable | |
100/Pk. |