Learn More
Fisherbrand™ Three-Sided Front Gamma Radiation Shield
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-S3G
Product Code. 12377187
  • £3910.00 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent shielding
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
Acrylic
Three sides front, Sides
Front: L x W: 50 x 46 cm (19.68 x 18.11 in.), Sides: 50 x 30 cm (19.68 x 11.81 in.)
Gamma radiation protection
Radiation shielded box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Three-Sided Front Gamma Radiation Shield > Three-Sided Front Gamma Radiation Shield