missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14318-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded byCD141 is an intronless gene is an endothelial-specific type I membrane receptor that binds thrombin. This binding results in the activation of protein C, which degrades clotting factors Va and VIIIa and reduces the amount of thrombin generated. Mutations in this gene are a cause of thromboembolic disease, also known as inherited thrombophilia.Specifications
Thrombomodulin | |
Polyclonal | |
Unconjugated | |
THBD | |
CD141, Fetomodulin, THBD, THRM, thrombomodulin, TM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7056 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07204 | |
THBD | |
Thrombomodulin Fusion Protein Ag5558 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |