missing translation for 'onlineSavingsMsg'
Learn More
Thulium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11365779

Thulium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11365779
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11365779

Brand: Thermo Scientific Alfa Aesar 010375.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-30-4
Molecular Formula Tm
Molecular Weight (g/mol) 168.93
MDL Number MFCD00011281
InChI Key FRNOGLGSGLTDKL-UHFFFAOYSA-N
Synonym tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
PubChem CID 23961
ChEBI CHEBI:33380
IUPAC Name thulium
SMILES [Tm]

Specifications

Color Silver to Gray
UN Number UN3178
Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈1.75g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Thulium foil, 0.25mm (0.01 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN08I019 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog Number010375.FF,010375.FI
Lot NumberN08I017 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog Number010375.FF,010375.FI
Lot NumberR28A018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010375.FF,010375.FI
Lot NumberF30U004 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010375.FF,010375.FI
Lot NumberB28T013 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010375.FF,010375.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.