missing translation for 'onlineSavingsMsg'
Learn More
Thulium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11306248

Thulium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11306248
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11306248

Brand: Thermo Scientific Alfa Aesar 011204.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thulium(III) fluoride is used for glass doping, and used to enhance mid-IR emission in oxyfluoride glass ceramics. It is also used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]

Specifications

Boiling Point 2200°C
Melting Point 1158°C
Physical Form Crystalline
Quantity 1 g
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 225.93
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 7.9 g/mL
Assay (REO)
Chemical Name or Material Thulium(III) fluoride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number22295 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011204.06,011204.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.