missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) oxide, REacton™, 99.99% (REO)

Product Code. 11306238
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11306238

Brand: Thermo Scientific Alfa Aesar 011199.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-44-1 | O3Tm2 | 385.87 g/mol

Thulium oxide is an excellent thulium source which finds use in glass, optical and ceramic applications. Further, it is used in the manufacturing of portable X-ray transmission device. In addition to this, it finds use as a nuclear reactor control material. Nanostructured thulium oxide acts as an efficient biosensor in the field of medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-44-1
Molecular Formula O3Tm2
Molecular Weight (g/mol) 385.87
MDL Number MFCD00011285
InChI Key ZIKATJAYWZUJPY-UHFFFAOYSA-N
IUPAC Name dithulium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Tm+3].[Tm+3]

Specifications

Melting Point 2425°C
Color Green
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (REO)
Solubility Information Slightly soluble in strong acids. Insoluble in water.
Formula Weight 385.87
Chemical Name or Material Thulium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.