Learn More
Chemical Identifiers
CAS | 65-71-4 |
---|---|
Molecular Formula | C5H6N2O2 |
Molecular Weight (g/mol) | 126.11 |
MDL Number | MFCD00006026 |
InChI Key | RWQNBRDOKXIBIV-UHFFFAOYSA-N |
Synonym | thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
PubChem CID | 1135 |
ChEBI | CHEBI:17821 |
IUPAC Name | 5-methyl-1H-pyrimidine-2,4-dione |
SMILES | CC1=CNC(=O)NC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10341184
|
Thermo Scientific Acros
157850050 |
5 g | Glass Bottle |
£19.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10478880
|
Thermo Scientific Acros
157850250 |
25 g | Plastic bottle |
£57.99
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704481
|
Thermo Scientific Acros
157851000 |
100 g | Plastic bottle |
£144.72
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11916521
|
Thermo Scientific Acros
157855000 |
500 g | Plastic bottle |
£489.44
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65-71-4 | |
126.11 | |
RWQNBRDOKXIBIV-UHFFFAOYSA-N | |
1135 | |
5-methyl-1H-pyrimidine-2,4-dione |
C5H6N2O2 | |
MFCD00006026 | |
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
CHEBI:17821 | |
CC1=CNC(=O)NC1=O |
Specifications
65-71-4 | |
White | |
98.5% min. (HPLC) | |
MFCD00006026 | |
15,9553 | |
Solubility in water: slightly soluble (4g/L 25°C). Other solubilities: soluble in hot water, readily soluble in alkalies, sparingly soluble in ether, practically insoluble in alcohol | |
CC1=CNC(=O)NC1=O | |
126.11 | |
CHEBI:17821 | |
99% | |
Thymine |
316.0°C to 317.0°C | |
Authentic | |
C5H6N2O2 | |
24,353 | |
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
RWQNBRDOKXIBIV-UHFFFAOYSA-N | |
5-methyl-1H-pyrimidine-2,4-dione | |
1135 | |
126.11 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-616-1
- Thymine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Thymine
RUO – Research Use Only