missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue Indicator 0.04% Alcoholic Solution, Reagecon
BIOPHARMA

High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.

Brand:  Reagecon TB04F


Product Code. 11513132

  • £42.05 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
  • This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications

Specifications

Accuracy ± 0.1%
HDPE Bottle GL45
Titration & Analyte Color Change
0.0004
500 mL
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • THYMOL BLUE 0.04% INDICATOR SOLUTION

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers