missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue, Pure, Indicator Grade, Fisher Chemical
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Specifications
Specifications
Melting Point | 221°C |
CAS | 76-61-9 |
Molecular Formula | C27H30O5S |
MDL Number | 5869 |
InChI Key | PRZSXZWFJHEZBJ-UHFFFAOYSA-N |
SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
IUPAC Name | 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
Molecular Weight (g/mol) | 466.592 |
PubChem CID | 65565 |
Formula Weight | 466.58g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'