missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

£21.81 - £68.51

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388957
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
038692.AE
100 mL
£21.81
100mL
In Stock
to see stock.
Add to basket
 
11430151
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
038692.AP
500 mL
£34.87
500mL
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
 
11398957
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
038692.M0
6 x 100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62625-21-2
488.57
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
C27H29NaO5S
MFCD00151093
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

62625-21-2
C27H29NaO5S
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
23692293
Odorless
Liquid
0.04% w/v aq. soln.
MFCD00151093
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
488.57
488.58
0.979 g/mL
Thymol Blue sodium salt
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-973-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only