missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein 0.05% Indicator Solution According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.

Brand:  Reagecon 1090701


Product Code. 11434749

  • £44.20 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • High quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
  • For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
  • A high-quality indicator solution such as Reagecon's facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications

Specifications

0.0005
Titration and Analyte Color Change
Amber Glass Winchester Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • THYMOLPHTHALEIN SOLUTION

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers