missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymolphthalein, ACS reagent
£32.60 - £564.00
Chemical Identifiers
CAS | 125-20-2 |
---|---|
Molecular Formula | C28H30O4 |
Molecular Weight (g/mol) | 430.53 |
InChI Key | LDKDGDIWEUUXSH-UHFFFAOYSA-N |
Synonym | thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one |
PubChem CID | 31316 |
IUPAC Name | 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10657581
|
Thermo Scientific Chemicals
420950100 |
10 g | Glass bottle |
£32.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10539292
|
Thermo Scientific Chemicals
420950250 |
25 g | Glass bottle |
£62.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10045930
|
Thermo Scientific Chemicals
420951000 |
100 g | Glass bottle |
£172.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11966191
|
Thermo Scientific Chemicals
420955000 |
500 g | Glass bottle |
£564.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
125-20-2 | |
430.53 | |
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
C28H30O4 | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
31316 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Specifications
125-20-2 | |
White to Yellow | |
C28H30O4 | |
15,9556 | |
Solubility in water: insoluble. Other solubilities: soluble in dilute alkali and acids, soluble in alcohol and acetone | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
430.53 | |
430.53 | |
ACS Reagent | |
Thymolphthalein |
251.0°C to 253.0°C | |
Authentic | |
18,I,381 | |
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one | |
31316 | |
passes test | |
Low Melting Mass |
Safety and Handling
missing translation for 'einecsNumber' : 204-729-7
RUO – Research Use Only