missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine Monoclonal Antibody (1H1), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA122092

Product Code. 11551340

  • £403.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%.

Thyroxine is a hormone produced by the thyroid glands to regulate metabolism by controlling the rate of oxidation in cells. Cases of hypothyroidism, where the gland is insufficiently active, can be treated by administration of thyroxine or a combination of thyroxine and triiodothyronine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thyroxine
Monoclonal
3.8 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Full length native protein, (purified) conjugated to BSA.
100 μg
Primary
4° C, do not freeze
Liquid
Chemical
ELISA
1H1
Unconjugated
Mouse
Protein A
RUO
Chemical, Mammalia
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.