missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroxine Monoclonal Antibody (XM212), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA122093
This item is not returnable.
View return policy
Description
This antibody reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%.
Thyroxine is a hormone produced by the thyroid glands to regulate metabolism by controlling the rate of oxidation in cells. Cases of hypothyroidism, where the gland is insufficiently active, can be treated by administration of thyroxine or a combination of thyroxine and triiodothyronine.Specifications
Thyroxine | |
Monoclonal | |
4.8 mg/mL | |
TBS with 0.05% sodium azide; pH 7.5 | |
Full length native protein (purified, Sigma T-2501) conjugated to BSA. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid | |
Chemical |
ELISA | |
XM212 | |
Unconjugated | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
Chemical, Mammalia | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction