missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thyroxine (T4) Competitive ELISA Kit

Product Code. p-200006748
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 15845863

Brand: Invitrogen™ EIAT4C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Thyroxine (T4) ELISA quantitates Thyroxine (T4) in serum, plasma, urine, dried fecal extracts, or cell culture medium. The assay will exclusively recognize both natural and recombinant Thyroxine (T4). Principle of the method The Thyroxine (T4) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Thyroxine is the main hormone produced by the thyroid gland. The thyroid hormones, triiodothyronine (T3) and thyroxine (T4), are tyrosine-based hormones produced by the thyroid gland that are primarily responsible for regulation of metabolism. Iodine is necessary for the production of T3 and T4. The major form of thyroid hormone in the blood is thyroxine, which has a longer half-life than T3. T4 is converted to the active T3 within cells by deiodinases (5’-iodinase). These are further processed by decarboxylation and deiodination to produce iodothyronamine and thyronamine. Thyroid hormone production is regulated via pituitary thyrotropin (TSH) modulation of thyroxine prohormone secretion by the thyroid gland and regulation of active triiodothyronine production. Thyroxine is identical across all species.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.625 to 20 ng/mL
Assay Sensitivity 0.29 ng/mL
Description Thyroxine (T4) Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Thyroid hormone
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 7%
Intraassay CV 3
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Metabolism
Sample Volume Fecal Extract 10 μL, Plasma 1 μL, Serum 1 μL, Supernatant 10 μL, Urine 2.5 μL
Storage Requirements -20°C
Target Thyroxine (T4)
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.