Learn More
TIAR Mouse anti-Human, Unconjugated, Clone: 1E8D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66907-1-IG-150UL
 View more versions of this product
Product Code. 16819195
  • £349.00 / 150µL
Explore more special offers
Description
Description
The TIAR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIAR. This antibody recognizes human, mouse, rat antigen. The TIAR antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
TIAR
Monoclonal
940 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1E8D12
Unconjugated
150 μL
7073
IgG1