Learn More
Manutan Tight head steel drum, lacquered, 30 litre capacity
Brand: Manutan A350867
This item is not returnable.
View return policy
Description
Suitable for the storage of liquids. All drums UN approved and have 1 x 51mm and 1 x 19mm tri-sure fitting.- Capacity : 30L
- Dimensions (H x D) : 572mm x 279mm
- UN specification : 1A1/Y