missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Mouse, PE, Clone: 1G9, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB57-PE-100ug

 View more versions of this product

Product Code. 15942228

  • £256.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TIGIT
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Tigit
Protein A/G Purified
RUO
100043314
IgG1 κ
Flow Cytometry
1G9
PE
XP_017172323
Mouse
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers