missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Mouse, PE, Clone: 1G9, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB57-PE-25ug
This item is not returnable.
View return policy
Specifications
TIGIT | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Tigit | |
Protein A/G Purified | |
RUO | |
100043314 | |
IgG1 κ |
Flow Cytometry | |
1G9 | |
PE | |
XP_017172323 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-4°C. |