missing translation for 'onlineSavingsMsg'
Learn More

Tiglic acid, 97+%

Product Code. 11431618
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11431618

Brand: Thermo Scientific Alfa Aesar A13905.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-59-1 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Melting Point 62°C to 66°C
Boiling Point 198°C to 199°C
UN Number UN3261
Beilstein 1236500
Merck Index 149433
Formula Weight 100.12
Percent Purity ≥98%
Density 0.969
Quantity 100 g
Flash Point 101°C (213°F)
Chemical Name or Material Tiglic acid
Show More Show Less
compliance-icons
Product Identifier
  • Tiglic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tiglic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.