missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM10B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10907-1-AP-150UL
This item is not returnable.
View return policy
Description
FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.Specifications
TIMM10B | |
Polyclonal | |
Unconjugated | |
TIMM10B | |
Fracture callus protein 1, FXC1, TIM10B, Tim9b, TIMM10B, TIMM9B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14356, 26515, 84384 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9WV96, Q9Y5J6 | |
Timm10b | |
TIMM10B Fusion Protein Ag1355 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction