Learn More
Timosaponin AIII, MedChemExpress
MedChemExpress Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Brand: Medchem Express HY-N0810
This item is not returnable.
View return policy
Description
Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.Specifications
Timosaponin AIII | |
White | |
C39H64O13 | |
DMSO : 125 mg/mL (168.71 mM; Need ultrasonic) | |
740.92 | |
98.0% | |
Solid |
41059-79-4 | |
Research | |
25 mg | |
C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@H](C)CO4)([H])C[C@@]1([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC2 | |
740.92 | |
Research | |
Cancer-programmed cell death |