missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP2 Mouse, Clone: SPM356, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7077-MSM1X-P1
This item is not returnable.
View return policy
Specifications
TIMP2 | |
Cytoplasm and Cell Surface | |
SPM356 | |
Unconjugated | |
NP_003246 | |
Mouse | |
21kDa | |
Protein A/G Purified | |
RUO | |
7077 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TIMP2 | |
A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI) | |
HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |