missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) acetate, 95%, Thermo Scientific Chemicals

£59.40 - £198.00

Chemical Identifiers

CAS 638-39-1
Molecular Formula C4H6O4Sn
Molecular Weight (g/mol) 236.798
MDL Number MFCD00054408
InChI Key PNOXNTGLSKTMQO-UHFFFAOYSA-L
Synonym tin ii acetate, stannous acetate, unii-1sku167w8p, tin 2+ diacetate, tin diacetate, tin acetate, tin di acetate, stannous diacetate, diacetoxytin, tech-95Show MoreShow Less
PubChem CID 69488
IUPAC Name tin(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328027
SDS View Documents
Thermo Scientific Alfa Aesar
022361.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11338027
SDS View Documents
Thermo Scientific Alfa Aesar
022361.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tin(II) acetate is used as a precursor in the production of tin anode, Sn-Cu bimetallic nanoparticles, high surface area tin oxide catalysts. It acts as Lewis acid catalysts for the esterification and transesterification reactions. It serves as an efficient catalytic system for carbon-hydrogen activation of methoxy group of anisole in presence of palladium(II) acetate. It plays an important role in stannous oral care compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 211-335-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Tin(II) acetate, 95%, Thermo Scientific Chemicals

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt