missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals Product Code.: 10772431

Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals

Product Code. 10772431
5 g
Click to view available options
Quantity:
5 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 10772431

Brand: Thermo Scientific Acros 196800050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
Molecular Weight (g/mol) 225.63
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate

Specifications

Boiling Point 652.0°C
Melting Point 37.0°C to 38.0°C
Color White or Colorless
Physical Form Adhering Crystalline Powder or Crystals
Quantity 5 g
Assay Percent Range 96% min. (Iodimetry)
Linear Formula SnCl2·2H2O
Fieser 01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68
Merck Index 15, 8910
Solubility Information Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Formula Weight 225.63
Percent Purity 97%
Packaging Glass Bottle
Density 2.7100g/mL
Chemical Name or Material Tin(II) chloride dihydrate, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0434606 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number196800050,196805000,196800025
Lot NumberA0431665 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number196800050,196805000,196800025
Lot NumberA0450045 Certificate TypeCertificate of Analysis Date07/02/2023 Catalog Number196800050,196805000,196800025
Lot NumberA0416834 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number196800050,196805000,196800025
Lot NumberA0268545 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number196800050,196805000,196800025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(II) chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.